Microplate Incubator
Compact and economical
  • Holds up to 14 microwell plates or 54 test tubes
  • Stack units to increase capacity without using more bench space
  • Heating element lies in the back-eliminating the risk of scorching the sample when placed on the chamber floor
$532.00 - $641.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (Liters) Control Type Power (VAC)
Availability Pricing
COMPARE
2.8 Analog 120
$532.00USD / Each
COMPARE
2.8 Analog 230
$641.00USD / Each