Micropump Compact Low-Flow Gear Pump Heads - C Mount Design
Offering remote C-mount choices for your low-flow applications
  • Use for applications requiring high initial differential pressure.
  • Suction-shoe design ensures complete fluid transfer for increased volumetric accuracy
  • Pump efficiency increases as discharge pressure increases
The compact pump head requires a drive for proper operation. Order drives separately.
Optional Service kit includes: gears, suction shoe, shaft seals, and springs. Order separately.
Contact our Application Specialists for assistance.
$1,198.00 - $1,359.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Differential Pressure (PSI) Flow Rate (mL/rev) Max Motor RPM (rpm)
Availability Pricing
Micropump Low-flow pump head, pressure-loaded, 0.092 mL/revolution EW-07002-12 Mfr # 81725 (GA-T23.PFS.C)
COMPARE
75 0.092 8000
$1,359.00USD / Each
Micropump C-Mount Cavity Style Pump Head; SS/PTFE/PTFE; 0.32 mL/rev EW-07002-20 Mfr # 82059 (GJ-N21.FF2S.C)
COMPARE
50 0.316 10000
$1,245.50USD / Each
Micropump Low-flow pump head, pressure-loaded, 0.017 mL/revolution EW-07002-32 Mfr # 82093 (GA-X21.CFS.C)
COMPARE
20 0.017 8000
$1,291.00USD / Each
Micropump Low-flow pump head, pressure-loaded, 0.042 mL/revolution EW-07002-33 Mfr # 82116 (GA-V21.CFS.C)
COMPARE
40 0.042 8000
$1,198.00USD / Each
Micropump Low-flow pump head, pressure-loaded, 0.084 mL/revolution EW-07002-35 Mfr # 82117 (GA-V23.CFS.C)
COMPARE
40 0.084 8000
$1,244.50USD / Each