Micropump® Eagle Drive™ Pump Systems
Complete pumps include head, motor, and integrated drive
  • Variable-speed electronic controller delivers smooth, pulseless flow
  • Static seals reduce maintenance
  • Small envelope-ideal for OEM use
Electromagnetically driven, precision-geared pumps provide smooth, accurate, pulseless flow. Torque improvements and compact size are achieved with NdFeB magnets. Control speed by using a 0 to 5 VDC input signal. Drives feature thermal and overload protection.
Choose a system with a suction shoe pump head to self compensate for wear, temperature, and pressure, and to maintain high volumetric efficiency. Cavity-style pump heads deliver higher lift capability and moderate pressure capability; an internal bypass is an integral safety feature of the cavity-style pump heads.
$2,166.00 - $2,392.50USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Flow Rate (mL/rev) Description
Availability Pricing
Micropump Gear Pump Systems, Suction Shoe Pump Head, 316SS/PPS/PTFE, 0-5 VDC EW-74215-10 Mfr # 85228
COMPARE
0.092 Gear Pump Systems, Suction Shoe Pump Head, 316SS/PPS/PTFE, 0-5 VDC
$2,392.50USD / Each
Micropump Gear Pump System, Cavity-style Pump Head, 316SS/PPS/PTFE, 0-5 VDC EW-74445-05 Mfr # 85462
COMPARE
0.32 Gear Pump System, Cavity-style Pump Head, 316SS/PPS/PTFE, 0-5 VDC
$2,221.50USD / Each
Micropump Gear Pump System, Cavity-style Pump Head, 316SS/PPS/PTFE, 0.64 ml/rev EW-74445-15 Mfr # 85463
COMPARE
0.64 Gear Pump System, Cavity-style Pump Head, 316SS/PPS/PTFE, 0.64 ml/rev
$2,166.00USD / Each
Micropump Gear Pump System, Cavity-style Pump Head, 316SS/PPS/PTFE, 0.91 ml/rev EW-74445-25 Mfr # 85464
COMPARE
0.91 Gear Pump System, Cavity-style Pump Head, 316SS/PPS/PTFE, 0.91 ml/rev
$2,313.00USD / Each