Mid-West Instruments Pulsation Dampeners
  • Provides infinitely adjustable dampening
  • Fine control needle valve for adjustable pressure pulsation dampening
  • Controlled leak check for protection against pressure spikes
  • Protects against surges and pressure shocks
  • 5000 (340 bar) or 10,000 (680 bar) PSIG working pressure
  • Optional swivel design eliminates gauge orientation problems
Model 150 is preferred over designs that feature several piston diameters or porous metal discs requiring removal and/or disassembly to readjust. The needle valve can be used as
a complete shutoff to facilitate changing out of a gauge or instrument. This method is not intended to replace instrument block valves. A continual over-torquing could damage the
valve seat. The 316 stainless steel ball check offers protection against surge and/or pressure spikes. The ball is driven on a seat by the pressure surge and held.
$38.50 - $126.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI)
Availability Pricing
Mid-West Instrument 150-BH-00 Pulsation Dampener, 1/2"Mx1/2"F, 3000 psi, Buna-N EW-68873-55 Mfr # 150-BH-00
COMPARE
3000
$48.25USD / Each
Mid-West Instrument 150-BO-00 Pulsation Dampener, 1/4"Mx1/4"F, 3000 psi, Buna-N EW-68873-57 Mfr # 150-BO-00
COMPARE
3000
$38.50USD / Each
Mid-West Instrument 150-BO-10 Pulsation Dampener, 1/4"Mx1/4"F, 3000 psi, Viton EW-68873-58 Mfr # 150-BO-10
COMPARE
3000
$42.75USD / Each
Mid-West Instrument 150-SH-00 Pulsation Dampener, 1/2"Mx1/2"F, 10000 psi, Buna-N EW-68873-59 Mfr # 150-SH-00
COMPARE
10000
$126.00USD / Each
Mid-West Instrument 150-SO-00 Pulsation Dampener, 1/4"Mx1/4"F, 5000 psi, Buna-N EW-68873-62 Mfr # 150-SO-00
COMPARE
5000
$94.00USD / Each
Mid-West Instrument 150-SO-10 Pulsation Dampener, 1/4"Mx1/4"F, 5000 psi, Viton EW-68873-63 Mfr # 150-SO-10
COMPARE
5000
$93.00USD / Each