Mini Needle Valves
Designed for industrial machinery, hydraulic, and pneumatic systems
  • Viton® O-ring seal and PTFE back-up ring protect from corrosion and galling
These compact valves are built for maximum durability and robust performance in the toughest applications, and are 100% helium leak-tested to 1 x 104 mL/s for guaranteed performance and reliability. They are available in brass, electroless nickel-plated steel, and electropolished 316 stainless steel. The body-to-bonnet, metal-to-metal seal is designed to significantly increase the pressure range of the valve without compromising the flow, and help maintain the integrity of the bonnet threads by segregating them from the process media.
The 100 Series hard-seat valves have a maximum pressure of 10,000 psi for stainless steel and steel models; 6000 psi for brass.
The 150 Series soft-tip valves feature a unique Delrin® acetal nonrotating soft tip stem, a backup metal-to-metal seal, and have a maximum pressure of 6000 psi for stainless steel and steel models; 3000 psi for brass.
$47.50 - $139.00USD / Each
59 variations of this product are available.
See All Product Options Below
59 PRODUCT OPTIONS
Compareitem specifications
Inlet Port Outlet Port Wetted Materials
Availability Pricing
NOSHOK 101MMB Hard Seat Mini Needle Valve 1/8", Straight Male-Male, Brass EW-08611-00 Mfr # 101MMB
COMPARE
1/8" NPT(M) 1/8" NPT(M) Brass
$52.00USD / Each
NOSHOK 101MMS Hard Seat Mini Needle Valve 1/8", Straight Male-Male, 316 ss EW-08611-08 Mfr # 101MMS
COMPARE
1/8" NPT(M) 1/8" NPT(M) 316 Stainless Steel
$111.00USD / Each
NOSHOK 102MMS Hard Seat Mini Needle Valve 1/4", Straight Male-Male, 316 ss EW-08611-10 Mfr # 102MMS
COMPARE
1/4" NPT(M) 1/4" NPT(M) 316 Stainless Steel
$111.00USD / Each
NOSHOK 102MFB Hard Seat Mini Needle Valve 1/4", Straight Male-Female, Brass EW-08611-14 Mfr # 102MFB
COMPARE
1/4" NPT(M) 1/4" NPT(F) Brass
$52.00USD / Each
NOSHOK 101MFC Hard Seat Mini Needle Valve 1/8", Straight Male-Female, Steel EW-08611-16 Mfr # 101MFC
COMPARE
1/8" NPT(M) 1/8" NPT(F) Steel
$47.50USD / Each
NOSHOK 102FFB Hard Seat Mini Needle Valve 1/4", Straight Female-Female, Brass EW-08611-26 Mfr # 102FFB
COMPARE
1/4" NPT(F) 1/4" NPT(F) Brass
$52.00USD / Each
NOSHOK 101FFC Hard Seat Mini Needle Valve 1/8", Straight Female-Female, Steel EW-08611-28 Mfr # 101FFC
COMPARE
1/8" NPT(F) 1/8" NPT(F) Steel
$47.50USD / Each
NOSHOK 102FFC Hard Seat Mini Needle Valve 1/4", Straight Female-Female, Steel EW-08611-30 Mfr # 102FFC
COMPARE
1/4" NPT(F) 1/4" NPT(F) Steel
$47.50USD / Each
NOSHOK 101MFAC Hard Seat Mini Needle Valve 1/8", 90 deg Angle, Male-Female, Steel EW-08611-40 Mfr # 101MFAC
COMPARE
1/8" NPT(M) 1/8" NPT(F) Steel
$59.50USD / Each
NOSHOK 101MFAS Hard Seat Mini Needle Valve 1/8", 90 deg Angle, Male-Female, 316 ss EW-08611-44 Mfr # 101MFAS
COMPARE
1/8" NPT(M) 1/8" NPT(F) 316 Stainless Steel
$139.00USD / Each
NOSHOK 102MFAS Hard Seat Mini Needle Valve 1/4", 90 deg Angle, Male-Female, 316 ss EW-08611-46 Mfr # 102MFAS
COMPARE
1/4" NPT(M) 1/4" NPT(F) 316 Stainless Steel
$139.00USD / Each
NOSHOK 101FFAC Hard Seat Mini Needle Valve 1/8", 90 deg Angle, Female-Female, Steel EW-08611-52 Mfr # 101FFAC
COMPARE
1/8" NPT(F) 1/8" NPT(F) Steel
$59.50USD / Each
NOSHOK 151MFB Soft Tip Mini Needle Valve 1/8", Straight Male-Female, Brass EW-08612-14 Mfr # 151MFB
COMPARE
1/8" NPT(M) 1/8" NPT(F) Brass
$52.00USD / Each
NOSHOK 152MFB Soft Tip Mini Needle Valve 1/4", Straight Male-Female, Brass EW-08612-16 Mfr # 152MFB
COMPARE
1/4" NPT(M) 1/4" NPT(F) Brass
$52.00USD / Each
NOSHOK 151MFS Soft Tip Mini Needle Valve 1/8", Straight Male-Female, 316 ss EW-08612-22 Mfr # 151MFS
COMPARE
1/8" NPT(M) 1/8" NPT(F) 316 Stainless Steel
$111.00USD / Each
NOSHOK 152MFAB Soft Tip Mini Needle Valve 1/4", 90 deg Angle, Male-Female, Brass EW-08612-40 Mfr # 152MFAB
COMPARE
1/4" NPT(M) 1/4" NPT(F) Brass
$64.50USD / Each
NOSHOK 152MFAC Soft Tip Mini Needle Valve 1/4", 90 deg Angle, Male-Female, Steel EW-08612-44 Mfr # 152MFAC
COMPARE
1/4" NPT(M) 1/4" NPT(F) Steel
$59.50USD / Each
NOSHOK 151MFAS Soft Tip Mini Needle Valve 1/8", 90 deg Angle, Male-Female, 316 ss EW-08612-46 Mfr # 151MFAS
COMPARE
1/8" NPT(M) 1/8" NPT(F) 316 Stainless Steel
$139.00USD / Each
NOSHOK 152MFAS Soft Tip Mini Needle Valve 1/4", 90 deg Angle, Male-Female, 316 ss EW-08612-48 Mfr # 152MFAS
COMPARE
1/4" NPT(M) 1/4" NPT(F) 316 Stainless Steel
$139.00USD / Each
NOSHOK 151FFAC Soft Tip Mini Needle Valve 1/8", 90 deg Angle, Female-Female, Steel EW-08612-54 Mfr # 151FFAC
COMPARE
1/8" NPT(F) 1/8" NPT(F) Steel
$59.50USD / Each
Showing 1 - 20 of 59
Need Help with this product?
Call Us At1-847-549-7600
Live chat