Mixed-Bed Deionization Cartridges
Deionization Cartridges are designed for deionizing water at 16 MΩ, the mixed-bed cartridges also minimize the TOC (total organic carbon) level. Liquid flows through a bed. Not a double or single open-end cartridge. Compatible with any 9-3/4" housing without a center rod (i.e. 01508-25 or VPN 150068). Not compatible with 01508-40 or 01508-41 (VPN 156017-02 or 156018-02).
All materials are FDA-compliant.
$60.50 - $237.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Capacity Filter Length (in) Flow Rate
Availability Pricing
Pentair 155273-03 Mixed-Bed deionization cartridge; 10"L, 0.25 GPM EW-29830-63 Mfr # 155273-43
COMPARE
270 grains CaCO3 10 0.25 GPM
$60.50USD / Each
Pentair 155274-03 Mixed-Bed deionization cartridge; 20"L, 0.5 GPM EW-29830-64 Mfr # 155274-03
COMPARE
600 grains CaCO3 20 0.5 GPM
$87.00USD / Each
Pentair 155281-43 Mixed-Bed deionization cartridge; 20"L, 1.25 GPM EW-29830-65 Mfr # 155281-43
COMPARE
1850 grains CaCO3 20 1.25 GPM
$237.00USD / Each