Economical Hand Mill
Economical hand mill for grinding soil, grains, or pharmaceuticals
Mill has large 50 cu. in. (820 cm3) capacity—grind up to 10 lb/hr to a maximum fineness of 75 to 100 mesh (approximately 150-µm). Rugged cast iron construction for maximum durability. Easily disassemble mill for cleaning without using tools. Clamp opens from 1/2" (1.3 cm) to 1-3/8" (3.5 cm); mill measures 14" (35.6 cm) H.
$372.50USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Feed Particle Size Sample Capacity (mL) Speed (rpm)
Availability Pricing
Hand Mill with Table Vise; 50 cu in Capacity, 150um Particle EW-04021-00 Mfr # S-F04BTB
(1)
COMPARE
0.125 in to 0.25 in
$372.50USD / Each