High-Flow Polypropylene Felt Filter Bags
Wide range of options for temperature, flow rate, and filtration quality
  • Constructed of Polypropylene Felt
Designed for higher flow with an average efficiency at the rated pore size. Polypropylene felt is ideal for general-purpose filtration. The bags are glazed to minimize fiber migration. Polypropylene felt filter bags are rated for 200°F (93°C) maximum.
$5.10USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Filter Media Flow Rate Micron Rating
Availability Pricing
COMPARE
Polyester 90 GPM 25
$5.10USD / Each