This product is now discontinued, View Related Products
Constant-Flow High-Pressure HPLC Gradient Piston Pump
Deliver precise volumes at pressures up to 6000 psi
  • Ideal for use in HPLC applications
  • Pulse dampener significantly reduces pulsation with low dead volume.
Use this dual head pump for analytical to small-scale chromatography applications. Constant-flow pump can be used with columns from 2 to 22 mm ID.
This dual-head drive features a linear cam and pressure compensation to provide accurate gradients in high- or low-pressure modes. Chemical-resistant keypad and digital LED display allow flow adjustments in 0.01-mL increments. Other features include RS-232 serial communication port, run/stop inputs, remote frequency and voltage controls, dual voltage capabilities, and real-time pressure monitoring.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (mL/min) Max Pressure (PSI) Wetted Materials
Availability Pricing
F05SFD01 Constant-Flow Gradient HPLC Piston Pump, 115/230 VAC EW-74930-10 Mfr # F05SFD01
COMPARE
12 6000 316 SS, fluorocarbon, ruby, sapphire
Call For Price
D05SFD01 Constant-Flow Gradient HPLC Piston Pump EW-74930-25 Mfr # D05SFD01
COMPARE
12 6000 316 SS, fluorocarbon, ruby, sapphire