Pail Mixer with Rim Clamp
Convenient for mixing materials in a closed 5-gallon bucket
  • Adjust motor along the centerline of the pail to align with the opening in lid
  • Folding impeller allows you to mount the mixer without removing the lid
  • Toggle switch on electric motor; air-control valve for speed adjustment on air-motor model
  • Air input pressure: 35 to 100 psi
$1,020.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Impeller Size (in) Motor hp Shaft Length (in)
Availability Pricing
Neptune Pail Mixer with Rim Clamp; 1/2- to 1-hp Air Drive EW-50500-30 Mfr # PAM-6.0
(1)
COMPARE
3.75 1/2 12
$1,020.00USD / Each