Dry Glass Bead Sterilizers
Effectively sterilize small, solid metal, and glass implements in seconds
  • Heated glass beads destroy all microbes and spores
  • Low power consumption allows for all day operation
  • Safe to use — no gases, fumes, open flames or harsh liquids
Sterilize solid metal and glass implements in less than ten seconds with these dry glass bead sterilizers. Glass beads are heated to 250°C to effectively destroy all microbes and spores. Suitable for sterilizing scissors, inoculating loops, and needles.
Sterilizers have a small footprint and can be conveniently placed on the benchtop. Power consumption is low allowing for all day operation. Dial thermometer on front of sterilizer ensures proper heating and back-up safety thermostat prevents over-heating.
$1,050.00 - $1,115.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Chamber Depth (in) Chamber Diameter (in) Power (VAC)
Availability Pricing
COMPARE
3 1.75 110
$1,050.00USD / Each
COMPARE
5 1.75 110
$1,110.00USD / Each
COMPARE
5 1.75 220
$1,115.00USD / Each