Gum Rubber Tubing, 25ft per pack
Applications: General-purpose liquid and gas applications
Characteristics Nontacky and seamless. Opaque amber.
Certifications: Tubing is manufactured according to GMP.
Temperature range: -15 to 158° F (-26 to 70° C)
Sterilize: Ethylene oxide.
$39.50 - $79.40USD / Pkg of 1
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
COMPARE
1/8 3/16 1/32
$39.50USD / Pkg of 1
COMPARE
1/8 1/4 1/16
$40.00USD / Pkg of 1
COMPARE
1/8 3/8 1/8
$62.80USD / Pkg of 1
COMPARE
3/16 5/16 1/16
$40.50USD / Pkg of 1
COMPARE
1/4 5/16 1/32
$41.50USD / Pkg of 1
COMPARE
1/4 3/8 1/16
$46.20USD / Pkg of 1
COMPARE
1/4 7/16 3/32
$58.70USD / Pkg of 1
COMPARE
1/4 1/2 1/8
$79.40USD / Pkg of 1
COMPARE
5/16 7/16 1/16
$51.40USD / Pkg of 1
COMPARE
5/16 1/2 3/32
$71.10USD / Pkg of 1
COMPARE
3/8 1/2 1/16
$63.80USD / Pkg of 1
Showing 1- 11 of 11