Miniature Gear Pumps - Compact Plastic Head and Drive
Complete mini gear pump systems for low-flow applications
  • Maximum flow rates up to 0.67 GPM
  • Choose from nonenclosed OEM, lab, and fully enclosed lab versions
  • 115 VAC, 230 VAC, and 12 VDC models
$76.70 - $220.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (GPM) Max Pressure (PSI) Description
Availability Pricing
COMPARE
0.43 10 Miniature gear pump, 0.43 GPM (1600 mL/min) non-enclosed OEM model, 115 VAC, 60 Hz
$107.00USD / Each
COMPARE
0.358 10 Miniature gear pump, 0.43 GPM (1600 mL/min) non-enclosed OEM model, 230 VAC, 50/60 Hz
$102.00USD / Each
COMPARE
0.67 21 Miniature Gear Pump, 0.67 GPM (2520 mL/min), 12 VDC Lab Model
$76.70USD / Each
COMPARE
0.32 10 Miniature gear pump, .32 GPM (1200 mL/min), fully enclosed lab model, 115 VAC, 60 Hz
$210.00USD / Each
COMPARE
0.267 10 Miniature gear pump, .32 GPM (1200 mL/min), 230 VAC, 50/60 Hz
$220.00USD / Each