This product is now discontinued, View Related Products
Bleach Spray System
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
  • Eliminates hazard of bleach spills on skin or surfaces
This two-bottle fixed-ratio trigger sprayer automatically mixes and dilutes 5.25% bleach concentrate with tap water to make a 5000 ppm bleach disinfectant solution for use on hard, non-porous surfaces. The sprayer keeps the bleach separate from the water until you spray, keeping the 10% solution fresh and effective. Refill the water cartridge with tap water and lock in the disposable bleach cartridges and system is ready for use. Item 18105-00 kit includes six, 11-oz. bleach cartridges, two, 5000 ppm sprayer heads, and two 11-oz. water bottles.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Applications Dilution Format
Availability Pricing
DEARDORFF FITZSIMMONS CORP 77062 Bleach Spray Bottle System Kit; 6 Cartridges and 2 Sprayer Heads/Bottles EW-18105-00 Mfr # 77062
COMPARE
Spray
Call For Price
DEARDORFF FITZSIMMONS CORP 77012 Bleach Refill, 12 11-oz. bleach cartridges EW-18105-02 Mfr # 77012
COMPARE
Spray
Call For Price
DEARDORFF FITZSIMMONS CORP 77006 Srayer with water bottles, 6/pk (six, 5000ppm sprayer heads with six, 11-oz. water bottles) EW-18105-04 Mfr # 77006
COMPARE
Spray