Portable LED Stroboscope
High-intensity light emitting diodes (LEDs) in a rugged all-metal design
  • Provides a long flash life - 5 times longer life then traditional xenon flashtubes
  • Ideal for motion and vibration analysis, and preventive maintenance programs
Easily monitor and inspect a variety of machine tools, components, raw or finished product, print media and many other applications under various speeds with this easy-to-use stroboscope. Unit features pulse control (adjustable pulse/focus) making it ideal for fine and gross viewing of fast-moving, fine movement details of small, high-speed moving objects like drill bits, fans and saw blades, micro motors, and more. The flash delay/phase shift control allows on-the-fly inspections of high-speed machinery and printing, without stopping the equipment, reducing critical downtime. Use the input/output connector port for internal or external LED flash triggering and for input external triggering sources such as remote sensors. Additional features include a simple manual dial, push button (X2), (/2), manual dial for flash rate controls, (+/–) control, and backlit LCD.
ITEM(S) ON SALE
$625.50USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Max Measuring Distance (in) Max RPM Range (rpm) Memory
Availability Pricing
Shimpo DT-326B Portable LED Stroboscope Kit EW-53565-75 Mfr # DT-326B
COMPARE
$625.50USD / Each
$695.00USD / Each