Seepex Progressing Cavity Pumps
Excellent chemical resistance, pulseless flow, and low shear
  • Easily move a variety of polymers and chemicals, and extremely adaptable to a wide range of industries
  • Gentle low-shear fluid transfer capable of pumping small particles without damaging product or pump
These pumps are of two main configurations: iron bodies with NBR stators and stainless steel (SS) bodies with Viton® stators. The two material combinations are ideal for many different industries including food & beverage, environmental, chemical & biochemical, pulp & paper, oil, gas & petrochemicals, agriculture, construction, biogas, ceramics, pharmaceutical and cosmetics . . . just to name a few!
The unique design of the rotor/stator combination provides many benefits—the smaller rotor cross section results in reduced thrust loads, the universal joint design offers improved NPSHR and low component wear, and the plug-in shaft connection allows for quick part change out. Listed below are the three most commonly asked-for drive configurations. The first is a standard AC motor (STD), the
second is a standard AC motor with an integrated controller to facilitate your changing requirements (IVD), and the third is the Spaggiari drive system (SPG).
$1,769.00 - $6,097.00USD / Each
47 variations of this product are available.
See All Product Options Below
47 PRODUCT OPTIONS
Compareitem specifications
Max Air Inlet Pressure (PSI) Max Flow Rate (GPH) Max Motor RPM (rpm)
Availability Pricing
Seepex MD00F Progressing Cavity Pump, HDPE/Titanium, IVD, 180 psi, 1.23-79 GPH EW-76803-08 Mfr # MD00F0BM120CEM05AC
COMPARE
79
$5,389.00USD / Each
Seepex W01NA1F0EM05AC Progressing Cavity Pump, Iron/Nitrile, IVD, 90 psi, 2.0 GPM EW-76803-17 Mfr # W01NA1F0LZ05AC
COMPARE
$3,818.00USD / Each
Seepex W01NA4H0BD1534 Progressing Cavity Pump, SS/Viton, STD, 90 psi, 5.5 GPM; AC EW-76803-19 Mfr # W01NA4H0WG1534
COMPARE
$2,746.50USD / Each
Seepex W01NA4H0EM05AC Progressing Cavity Pump, SS/Viton, IVD, 90 psi, 2.0 GPM EW-76803-21 Mfr # W01NA4H0LZ05AC
COMPARE
$4,629.50USD / Each
Seepex W02NA1F0BD1534 Progressing Cavity Pump, Iron/Nitrile, STD, 60 psi, 10.5 GPM; AC EW-76803-25 Mfr # W02NA1F0WG1534
COMPARE
$1,873.00USD / Each
Seepex W02NA4H0BD1534 Progressing Cavity Pump, SS/Viton, STD, 60 psi, 10.5 GPM; AC EW-76803-29 Mfr # W02NA4H0WG1534
COMPARE
$2,746.50USD / Each
Seepex W05NA4H0BD2036 Progressing Cavity Pump, SS/Viton, STD, 45 psi, 29.5 GPM; AC EW-76803-37 Mfr # W05NA4H0WG2036
COMPARE
$4,099.00USD / Each
Seepex W05NA4H0BD3034 Progressing Cavity Pump, SS/Viton, STD, 45 psi, 46 GPM; AC EW-76803-38 Mfr # W05NA4H0WG2036
COMPARE
$3,683.00USD / Each
Seepex W10NA1F0BD5036 Progressing Cavity Pump, Iron/Nitrile, STD, 45 psi, 62.5 GPM; AC EW-76803-41 Mfr # W10NA1F0WG5036
COMPARE
$5,098.00USD / Each
Seepex W01NA1F0BD1036 Progressing Cavity Pump, Iron/Nitrile, STD, 90 psi, 3.7 GPM; AC EW-76803-46 Mfr # W01NA1F0WG1036
COMPARE
$2,018.50USD / Each
Seepex W01NA4H0BD1036 Progressing Cavity Pump, SS/Viton, STD, 90 psi, 3.7 GPM; AC EW-76803-61 Mfr # W01NA4H0WG1036
COMPARE
$2,892.50USD / Each
Seepex W02NA1F0BD1036 Progressing Cavity Pump, Iron/Nitrile, STD, 60 psi, 7.0 GPM; AC EW-76803-63 Mfr # W02NA1F0WG1036
COMPARE
$2,018.50USD / Each
Seepex W02NA1F0BD1514 Progressing Cavity Pump, Iron/Nitrile, STD, 60 psi, 10.5 GPM; AC EW-76803-64 Mfr # W02NA1F0WG1514
COMPARE
$1,769.00USD / Each
Seepex W02NA1H0BD1036 Progressing Cavity Pump, Iron/Viton, STD, 60 psi, 7.0 GPM; AC EW-76803-66 Mfr # W02NA1H0WG1036
COMPARE
$2,164.00USD / Each
Seepex W02NA1H0BD1514 Progressing Cavity Pump, Iron/Viton, STD, 60 psi, 10.5 GPM; AC EW-76803-67 Mfr # W02NA1H0WG1514
COMPARE
$1,914.50USD / Each
Seepex W02NA4F0BD1036 Progressing Cavity Pump, SS/Viton, STD, 60 psi, 7.0 GPM; AC EW-76803-71 Mfr # W02NA4F0WG1036
COMPARE
$2,746.50USD / Each
Seepex W05NA1H0BD2036 Progressing Cavity Pump, Iron/Viton, STD, 45 psi, 29.5 GPM EW-76803-81 Mfr # W05NA1H0WG2036
COMPARE
$3,880.50USD / Each
Seepex W05NA1H0BD3034 Progressing Cavity Pump, Iron/Viton, STD, 45 psi, 46 GPM EW-76803-82 Mfr # W05NA1H0WG3034
COMPARE
$3,464.50USD / Each
Seepex W05NA4F0BD3034 Progressing Cavity Pump, SS/Nitrile, STD, 45 psi, 46.0 GPM EW-76803-86 Mfr # W05NA4F0WG3034
COMPARE
$3,235.50USD / Each
Seepex W10NA4F0BD5036 Progressing Cavity Pump, SS/Nitrile, STD, 45 psi, 62.5 GPM EW-76803-91 Mfr # W10NA4F0WG5036
COMPARE
$5,504.00USD / Each
Showing 1 - 20 of 47