Pressure Dataloggers with LCD Display
Monitor pressure up to 500psi at-a-glance or with one-step data retrieval
  • USB-enabled triple speed downloading
  • 1-second sampling rate
  • IP68 case
  • FLASH card option
  • User replaceable battery
  • Pressure sensor includes built in diaphragm seal
One-step data retrieval from datalogger to a FLASH memory card makes the routine of gathering data fast and easy. By pressing a single button, all your data will be downloaded to the removable memory card (optional), thereby eliminating the need to bring a laptop or move the datalogger. A Card Reader Kit, 23036-52, and a memory card, 23036-53, are all you need for this data retrieval system.
Chose from 0 to 100 psi, 0-300 psi, or 0 to 500 psi models. All models feature LCD display with min/max option, unit of measure indicator, low battery indicator, and much more. An optional locking case is available to prevent tampering.
$24.00 - $730.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Chart Diameter (in) Chart Speed Max Input (VDC)
Availability Pricing
Dickson A721 Locking Case for PR125/325/525 Loggers EW-68511-35 Mfr # A721
COMPARE
$125.00USD / Each
Dickson A125 3.0 V Lithium Battery, CR2450; 1/Pk EW-68511-36 Mfr # A124
COMPARE
$24.00USD / Each
Dickson PR125 Pressure logger with display, 0 to 100 PSI EW-68512-04 Mfr # PR125
COMPARE
$730.00USD / Each
Dickson PR325 Pressure logger with display, 0 to 300 PSI EW-68512-05 Mfr # PR325
COMPARE
$730.00USD / Each