Modular Liquid Flow Rate Sensors
System design is flexible to suit a wide range of applications
  • Brass sensors for high pressure; PTFE sensors for high-purity systems; Ryton® PPS sensors for aggressive liquids
  • Interface with data acquisition systems
  • Dual signal outputs to monitor and record flow rates and totals
Use these modular flow sensors and totalizers for any type of process or laboratory application. The flow sensors measure low viscosity liquids from 13 mL/min up to 10 L/min. For a complete system, order a flow sensor, power supply, and flow display/totalizer.
These sensors use an optical beam, turbine wheel, and photodiode translator to generate a 7.5 V pulse
output that is proportional to the flow rate. Sensors also provide a highly linear 0 to 5 VDC output signal
for control or data logging purposes. Note: Due to the optical pick-up, the sensors should not be used with opaque liquids. Ryton® PPS sensors have wetted materials of polyphenylene sulfide (PPS), glass, sapphire, and Viton®. Brass sensors have wetted materials of brass, glass, and Viton. PTFE sensors have wetted materials of PTFE, sapphire, and Kalrez. A separate power supply is required for these flow sensors—(order separately). If a Ryton® PPS or brass sensor is being used with an existing power supply, order cable assembly 32704-52 (PTFE models already include their own cable assembly).
$399.00 - $807.00USD / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (mL/min) Min Flow Rate (mL/min) Wetted Materials
Availability Pricing
McMillan Flow Modular PTFE Rate Sensor for Liquids, 100 to 1000 mL/min (Reconditioned)
Steals and Deals
EW-00133-BN Mfr # 32705-04 RECONDITIONED
COMPARE
Call For Price
McMillan Flow 101-3T Modular Ryton PPS Flow Rate Sensor for Liquids, 13 to 100 mL/min EW-32704-00 Mfr # 101-3T
COMPARE
100 13 PPS, Glass, Sapphire, and Viton
$399.00USD / Each
McMillan Flow 101-5T Modular Ryton PPS Flow Rate Sensor for Liquids, 50 to 500 mL/min EW-32704-02 Mfr # 101-5T
COMPARE
500 50 PPS, Glass, Sapphire, and Viton
$401.00USD / Each
McMillan Flow 101-6T Modular Ryton PPS Flow Rate Sensor for Liquids, 100 to 1000 mL/min EW-32704-04 Mfr # 101-6T
COMPARE
1000 100 PPS, Glass, Sapphire, and Viton
$401.00USD / Each
McMillan Flow 101-7T Modular Ryton PPS Flow Rate Sensor for Liquids, 0.1 to 2 LPM EW-32704-06 Mfr # 101-7T
COMPARE
2000 200 PPS, Glass, Sapphire, and Viton
$399.00USD / Each
McMillan Flow 101-8T Modular Ryton PPS Flow Rate Sensor for Liquids, 0.2 to 5 LPM EW-32704-08 Mfr # 101-8T
COMPARE
5000 500 PPS, Glass, Sapphire, and Viton
$470.00USD / Each
McMillan Flow 101-9T Modular Ryton PPS Flow Rate Sensor for Liquids, 1 to 10 LPM EW-32704-10 Mfr # 101-9T
COMPARE
10000 1000 PPS, Glass, Sapphire, and Viton
$561.00USD / Each
McMillan Flow 102-3T Modular Brass Flow Rate Sensor for Liquids, 13 to 100 mL/min EW-32704-12 Mfr # 102-3T
COMPARE
100 13 Brass, Glass, Viton
$646.00USD / Each
McMillan Flow 102-5T Modular Brass Flow Rate Sensor for Liquids, 50 to 500 mL/min EW-32704-14 Mfr # 102-5T
COMPARE
500 50 Brass, Glass, Viton
$646.00USD / Each
McMillan Flow 102-6T Modular Brass Flow Rate Sensor for Liquids, 100 to 1000 mL/min EW-32704-16 Mfr # 102-6T
COMPARE
1000 100 Brass, Glass, Viton
$646.00USD / Each
McMillan Flow 102-7T Modular Brass Flow Rate Sensor for Liquids, 0.1 to 2 LPM EW-32704-18 Mfr # 102-7T
COMPARE
2000 200 Brass, Glass, Viton
$646.00USD / Each
McMillan Flow 102-8T Modular Brass Flow Rate Sensor for Liquids, 0.2 to 5 LPM EW-32704-20 Mfr # 102-8T
COMPARE
5000 500 Brass, Glass, Viton
$719.00USD / Each
McMillan Flow 102-9T Modular Brass Flow Rate Sensor for Liquids, 1 to 10 LPM EW-32704-22 Mfr # 102-9T
COMPARE
10000 1000 Brass, Glass, Viton
$807.00USD / Each
Showing 1- 13 of 13