Motic Advanced Metallurgical Microscopes
Inspect opaque materials with ease and affordability
  • Hard-coated and chemical-resistant stages withstand harsh environments
  • Plan achromatic objectives feature the color-corrected infinity optical system (CCIS®) for improved image quality
  • Power supply located away from the stand to avoid subjecting the microscope to unnecessary heat
  • Push polarizer into place and turn the analyzer thumbwheel to effortlessly optimize image contrast
  • Adjustable interpupillary distance and ergonomic viewing angle reduce fatigue
This series of robust incident light microscopes features four objectives, with one available socket in the nosepiece for a supplementary objective. Eyepieces are treated with an antifungal agent to ensure a long lifetime, and have a high eyepoint for eyeglass wearers. For added security, the eyepieces can be fixed to the microscope body by locking screws.
$4,250.50 - $5,152.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Head Configuration Maximum Magnification Stage Dimensions
Availability Pricing
Motic Instruments BA310MET Advanced Binocular Metallurgical Microscope EW-48405-01 Mfr # 1100100402144
COMPARE
Binocular 500x 180 x 140 mm
$4,250.50USD / Each
Motic Instruments BA310MET Advanced Trinocular Metallurgical Microscope EW-48405-03 Mfr # 1100100402045
COMPARE
Trinocular 500x 180 x 140 mm
$4,470.50USD / Each
Motic Instruments BA310MET-T Advanced Trinocular Metallurgical Microscope, 3" x 2" light-transmitting stage EW-48405-07 Mfr # 1100100402601
COMPARE
Trinocular 500x 240 x 140 mm
$4,829.00USD / Each
Motic Instruments BA310MET-T Advanced Trinocular Metallurgical Microscope, 6" x 4" light-transmitting stage EW-48405-09 Mfr # 1100100402524
COMPARE
Trinocular 500x 300 x 180 mm
$5,152.00USD / Each