Multi-Channel Tubing Racks, Non-Adhesive
An easy solution to secure and route multiple tubing lines
  • Keep your tubing lines organized and untangled
These tube racks offer a simple and economical method for organizing your tubing lines. The 3/16" and 1/4" sizes can be easily separated down to single holders by hand for increased flexibility and custom tubing arrangements. The 3/16" and 1/4" sizes are available with adhesive fastener, and all sizes are available with through holes for secure mounting (mounting screws are not included). Standard racks are black polyethylene; 3/16" and 1/4" sizes are available in other colors. Contact Cole-Parmer for more information.
$20.50 - $88.50USD / Pkg of 10
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
For Tubing OD (in) Number of Channels
Availability Pricing
COMPARE
1/4 10
$20.50USD / Pkg of 10
COMPARE
3/8 7
$88.50USD / Pkg of 10
COMPARE
1/2 5
$57.00USD / Pkg of 10
COMPARE
3/16 10
$26.25USD / Pkg of 10
COMPARE
1/4 10
$30.75USD / Pkg of 10