Multi-Tube Connectors
Save time -- with one coupling you can connect two, six, or ten lines
Colder has made it easy to handle multiple tubing lines in your process that require connection and reconnection! These multiline couplings simplify the manual steps in your process for quick transfers and minimize your down time. Twin Tube™ series: With two lines in one fitting, you can connect or disconnect with the push of a button. Each line is capable of independently handling anything from vacuum to 120 psi. The coupling allows rotation of both tubing lines to prevent kinking. The Twin Tube™ series is available with hose barb connections only. Sixtube™ series: With a single connector, control six different lines! An efficient thumblatch design allows quick disconnecting with optional shutoff.
$8.40 - $600.00USD / Each
51 variations of this product are available.
See All Product Options Below
51 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Fitting Material
Availability Pricing
CPC (Colder) PTC16020 Coupling Body, Two-Line, Acetal, 1/8" ID Tube EW-31050-02 Mfr # PTC16020
COMPARE
1/8 Acetal
$16.00USD / Each
CPC (Colder) PTC1602096 Coupling Body, Two-Line, ABS, 1/8" ID Tube EW-31050-03 Mfr # PTC1602096
COMPARE
1/8 ABS Plastic
$12.70USD / Each
CPC (Colder) PTC22020 Coupling Insert, Two-Line, Acetal, 1/8" ID Tube EW-31050-05 Mfr # PTC22020
COMPARE
1/8 Acetal
$8.90USD / Each
CPC (Colder) SXM17M3 Coupling Body, Six-Line, Male, Acetal, 3mm ID Tube; 10/PK EW-31307-74 Mfr # SXM17M3
COMPARE
Acetal
$210.00USD / Pkg of 10
CPC (Colder) SXF1702 Coupling Body, Six-Line, Female, Acetal, 1/8" ID Tube; 10/PK EW-31307-78 Mfr # SXF1702
COMPARE
1/8 Acetal
$157.00USD / Pkg of 10
CPC (Colder) SXFD17M3 Coupling Body, Six-Line, Female, Valved, Acetal, 3mm ID Tube; 10/PK EW-31307-80 Mfr # SXFD17M3
COMPARE
Acetal
$600.00USD / Pkg of 10
CPC (Colder) SXF170112 Coupling Body, Six-Line, Female, Polypropylene, 1/16" ID Tube; 10/PK EW-31307-83 Mfr # SXF170112
COMPARE
1/16 Polypropylene
$202.00USD / Pkg of 10
CPC (Colder) SXM42M3 Coupling Insert, Six-Line, Male, Acetal, 3mm ID Tube; 10/PK EW-31307-85 Mfr # SXM42M3
COMPARE
Acetal
$130.00USD / Pkg of 10
CPC (Colder) SXF42M3 Coupling Insert, Six-Line, Female, Acetal, 3mm ID Tube; 10/PK EW-31307-87 Mfr # SXF42M3
COMPARE
Acetal
$135.00USD / Pkg of 10
CPC (Colder) TT1001 Complete Coupling, Ten-Line, Acetal, 1/16" ID Tube; 5/PK EW-31307-96 Mfr # TT1001
COMPARE
1/16 Acetal
$210.00USD / Pkg of 5
CPC (Colder) TT10M3 Complete Coupling, Ten-Line, Acetal, 3mm ID Tube; 5/PK EW-31307-97 Mfr # TT10M3
COMPARE
Acetal
$250.00USD / Pkg of 5
CPC (Colder) TT1002 Complete Coupling, Ten-Line, Acetal, 1/8" ID Tube; 5/PK EW-31307-98 Mfr # TT1002
COMPARE
1/8 Acetal
$210.00USD / Pkg of 5
CPC (Colder) TTD1001 Complete Coupling, Ten-Line, Valved, Acetal, 1/16" ID Tube; 5/PK EW-31307-99 Mfr # TTD1001
COMPARE
1/16 Acetal
$558.00USD / Pkg of 5
CPC (Colder) TT100212 Complete Coupling, Ten-Line, Polypropylene, 1/8" ID Tube; 5/PK EW-31308-06 Mfr # TT100212
COMPARE
1/8 Polypropylene
$273.00USD / Pkg of 5
CPC (Colder) TFB10M3 Coupling Body, Ten-Line, Acetal, 3mm ID Tube; 5/PK EW-31308-08 Mfr # TFB10M3
COMPARE
Acetal
$74.50USD / Pkg of 5
CPC (Colder) TFBD1001 Coupling Body, Ten-Line, Valved, Acetal, 1/16" ID Tube; 5/PK EW-31308-14 Mfr # TFBD1001
COMPARE
1/16 Acetal
$414.00USD / Pkg of 5
CPC (Colder) TFBD10M3 Coupling Body, Ten-Line, Valved, Acetal, 3mm ID Tube; 5/PK EW-31308-15 Mfr # TFBD10M3
COMPARE
Acetal
$445.00USD / Pkg of 5
CPC (Colder) TMB1001 Coupling Insert, Ten-Line, Acetal, 1/16" ID Tube; 5/PK EW-31308-19 Mfr # TMB1001
COMPARE
1/16 Acetal
$131.00USD / Pkg of 5
CPC (Colder) TMB10M3 Coupling Insert, Ten-Line, Acetal, 3mm ID Tube; 5/PK EW-31308-24 Mfr # TMB10M3
COMPARE
Acetal
$146.00USD / Pkg of 5
CPC (Colder) TMB100212 Coupling Insert, Ten-Line, Polypropylene, 1/8" ID Tube; 5/PK EW-31308-27 Mfr # TMB100212
COMPARE
1/8 Polypropylene
$176.00USD / Pkg of 5
Showing 1 - 20 of 51