This product is now discontinued, View Related Products
Multifunction Portable Calibrators
Highly Accurate Field and Lab Measurement Models
  • Measure & generate voltage, current & resistance signals
  • Scaling, steps, synthesizer, square root, statistical functions make advanced data exploitation easier
  • Quick connect terminals for Banana Plugs, TC connectors or bare wires (to 3mm in diameter)
  • Rugged IP54 construction with anti-shock boot
All calibrator models feature an ergonomic design and embedded software allowing your to integrate all your required calibration functionality for a broad range of industry process instrument calibrations into a single unit. All models are designed with a large dual display which makes measure and sourcing in the field easy to read and make adjustments. Choose from four models with or without pressure, average to high-level accuracy, and with or without memory for storage of calibration readings which can be downloaded into a PC for analysis and to issue calibration certificates.
Features of all Models
oSimultaneous measurement & generation on isolated channels
oTemperature by Resistive Probes & Thermocouples
oFrequency for frequency signals & for dry contacts
oUSB connection/Rechargeable NiMH Batteries
Additional Features of C75 Model
oPressure measurements when used with optional external pressure modules
oSwitch Test for temperature or pressure
oMemory capacity of 10,000 records
oCompatible with optional DataCal-Lite or 21 CFR Part 11, FDA compliant DataCal software
Additional Features of C100 Model
oSame as C75 but with higher precision Laboratory-Grade accuracy (specifications available)
Additional features of C150 Model
oHighest accuracy available - 0.005% over 1 year (specifications available)
oFull compliance with NADCAP heat treatment standard
oCalibration of Thermistors
oCalibration of HART transmitters (includes HART compatible software)
oComparison calibration with dual input channels (In/Out or In/In)
oBlue Tooth Interface
Call For Price
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
Compareitem specifications
Max Measurement (mA) Max Measurement (VDC)
Availability Pricing
COMPARE
Call For Price
COMPARE
Call For Price
Palmer Wahl C100 Portable multifunction high-accuracy calibrator, with pressure and memory EW-50003-03 Mfr # C100
COMPARE
Call For Price
Palmer Wahl C150 Wahl Dual-Channel High-Accuracy Portable Multifunction Calibrator EW-50003-04 Mfr # C150
COMPARE
Call For Price
COMPARE
Call For Price
COMPARE
Call For Price
COMPARE
Call For Price
COMPARE
Call For Price
Palmer Wahl PMA0001 Wahl Absolute Pressure Module, 14.5 psia EW-50003-51 Mfr # PMA0001
COMPARE
Call For Price
Palmer Wahl PMA0003 Wahl Absolute Pressure Module, 43.5 psia EW-50003-52 Mfr # PMA0003
COMPARE
Call For Price
Palmer Wahl PMA0010 Wahl Absolute Pressure Module, 145 psia EW-50003-53 Mfr # PMA0010
COMPARE
Call For Price
Palmer Wahl PMA0030 Wahl Absolute Pressure Module, 435 psia EW-50003-54 Mfr # PMA0030
COMPARE
Call For Price
Palmer Wahl PMA0100 Wahl Absolute Pressure Module, 1450 psia EW-50003-55 Mfr # PMA0100
COMPARE
Call For Price
Palmer Wahl PMA0300 Wahl Absolute Pressure Module, 4350 psia EW-50003-56 Mfr # PMA0300
COMPARE
Call For Price
Palmer Wahl PMA1000 Wahl Absolute Pressure Module; 14,500 psia EW-50003-57 Mfr # PMA1000
COMPARE
Call For Price
Palmer Wahl PMR0001 Wahl Relative Pressure Module, 14.5 psi EW-50003-58 Mfr # PMR0001
COMPARE
Call For Price
Palmer Wahl PMR0003 Wahl Relative Pressure Module, 43.5 psi EW-50003-59 Mfr # PMR0003
COMPARE
Call For Price
Palmer Wahl PMR0010 Wahl Relative Pressure Module, 145 psi EW-50003-60 Mfr # PMR0010
COMPARE
Call For Price
Palmer Wahl PMR0030 Wahl Relative Pressure Module, 435 psi EW-50003-61 Mfr # PMR0030
COMPARE
Call For Price
Showing 1- 19 of 19