Mutimode Electronic Test Indicator
Fowler's QuadraTest offers unrivaled measuring versatility in an Electronic Test Indicator.
  • 4 measuring modes: Min, Max, TIR, and Direct
  • True inch / metric conversion
  • Water, oil and coolant resistant to IP65
  • 1-1/2" diameter dial face rotates a full 360 °
  • Bar graph and numeric digital display
  • Meets or exceeds DIN /ISO standards
  • Maximum error on .0005" scale at full range: .0004"
  • .080" contact ball diameter
$236.50 - $525.75USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Fowler 54-562-777 1/2" Electronic Test Indicator (Depth Gauges/Indicators) EW-10508-52 Mfr # 54-562-777
COMPARE
1/2" Electronic Test Indicator (Depth Gauges/Indicators)
$453.00USD / Each
Fowler 54-562-888 1 7/16" Electronic Test Indicator (Depth Gauges/Indicators) EW-10508-53 Mfr # 54-562-888
COMPARE
1 7/16" Electronic Test Indicator (Depth Gauges/Indicators)
$525.75USD / Each
Fowler 54-115-444 Mutimode Electronic Test Indicator Output Cable EW-10508-63 Mfr # 54-115-444
COMPARE
Mutimode Electronic Test Indicator Output Cable
$236.50USD / Each