This product is now discontinued, View Related Products

N-Bromosuccinimide 36893

Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Acros Organics AC107450010 N-Bromosuccinimide 99% (1kg) EW-88096-25 Mfr # AC107450010
COMPARE
128-08-5
Call For Price
Acros Organics AC107450100 N-Bromosuccinimide 99% (10g) EW-88096-26 Mfr # AC107450100
COMPARE
128-08-5
Call For Price
Acros Organics AC107451000 N-Bromosuccinimide 99% (100g) CAS 128-08-5 EW-88096-27 Mfr # AC107451000
COMPARE
128085
Call For Price
Acros Organics AC107455000 N-Bromosuccinimide 99% (500g) EW-88096-28 Mfr # AC107455000
COMPARE
128-08-5
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat