n-Butanol

  • Assay - ≥99.8%
  • Color (APHA) - ≤5
  • Isobutanol - ≤0.1%
  • Water - ≤0.1%
  • Acidity - ≤0.005%
  • Aldehydes - None
  • Chlorides - None
$57.50 - $238.00USD / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
CAS Number Volume (mL) Description
Availability Pricing
Fisher BioReagents BP50525 n-Butanol (Clear, Colorless Liq.) (25 mL) EW-88067-49 Mfr # BP50525
COMPARE
71363 25 n-Butanol (Clear, Colorless Liq.) (25 mL)
$57.50USD / Each
Fisher BioReagents BP505500 n-Butanol (Clear, Colorless Liq.) (500 mL) EW-88067-50 Mfr # BP505500
COMPARE
71363 500 n-Butanol (Clear, Colorless Liq.) (500 mL)
$238.00USD / Each
COMPARE
71-36-3 1000 n-Butyl Alcohol, ACS Grade, 1 L
$72.00USD / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat