This product is now discontinued, View Related Products

n-Butylamine

  • Assay (w/w) - 98-100%
  • Color (APHA) - ≤15
  • Water - ≤2.0%
Call For Price

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Fisher Chemical B415500 n-Butylamine (Certified) (500mL) EW-88056-03 Mfr # B415500
COMPARE
109-73-9
Call For Price
Acros Organics AC107800010 n-Butylamine 99.5% (1l) EW-88096-91 Mfr # AC107800010
COMPARE
109-73-9
Call For Price
Acros Organics AC107800025 n-Butylamine 99.5% (2.5l) EW-88096-92 Mfr # AC107800025
COMPARE
109-73-9
Call For Price
Acros Organics AC107800050 n-Butylamine 99.5% (5ml) EW-88096-93 Mfr # AC107800050
COMPARE
109-73-9
Call For Price
Acros Organics AC219740010 n-Butylamine p.a. (1l) EW-88235-07 Mfr # AC219740010
COMPARE
109-73-9
Call For Price
Acros Organics AC219742500 n-Butylamine p.a. (250ml) EW-88235-08 Mfr # AC219742500
COMPARE
109-73-9
Call For Price
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat