N-TRON Industrial Ethernet Switch 59714
The N-Tron 104TX / 105TX is a low cost unmanaged industrial ethernet switch. It is housed in a ruggidized DIN-rail enclosure and is designed for use in mission critical data acquisition, controland Ethernet I/O applications.
  • Compact, space saving package
  • Full IEEE 802.3 compliance
  • Four 10/100 Base TX FJ-45 Ports (140TX) / Five 10/100 Base TX RJ-45 Ports (105TX)
  • Unmanaged operation
  • Extended environmental specifications:
    • -40C to 80C operating temperature
    • ->2M hours MTBF
  • Supports full/half duplex operation
  • Up to 1.0 Gb/s maximum throughput
  • MDIX auto sensing cable
  • Auto sensing speed and flow control
  • Full wire speed communications
  • Store and forward technology
  • Redundant Power Inputs (10-30 VDC)
  • LED link/activity status indication
  • Hardened metal DIN-Rail enclosure
$121.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Max Input Current (mA) Max Input Voltage (VDC) Number of Ports
Availability Pricing
Red Lion 105TX Ethernet Switch, Five 10/100BaseTX RJ-45 Port EW-26047-23 Mfr # 105TX
COMPARE
215 30 5
$121.00USD / Each