This product is now discontinued, View Related Products
N-TRON Industrial PoE Mid-Span Injector
Get PoE (Power over Ethernet) capability without the need for costly electrical wiring
  • Four 10/100BaseTX RJ-45 ports (data in)
  • Four 10/100BaseTX RJ-45 PoE ports (data and power out)
  • Unmanaged operation
  • Automatic detection of connected PoE devices
  • Auto-disconnect of PoE port if fault is detected
  • Supports 15.4 watts per port (13 watts at the PD)
  • Full wire speed communications
  • Up to 0.8 Gb/s maximum throughput
  • Power fault status LED
  • LED PoE status indication
  • Redundant power inputs (46 to 49 VDC)
  • ESD protection diodes on RJ-45 ports
  • Surge protection diodes on power inputs
  • Hardened metal DIN-rail enclosure
  • Extended environmental specifications
  • – Operating temperature of –40 to 85°C
  • – >2M hours MTBF
  • Compact, space-saving design
  • IEEE 802.3af compliance
Ideal for upgrading non-PoE network infrastructures with PoE capabilities, the 100-POE4 Industrial PoE Mid-Span Injector is designed to solve the most demanding industrial communications requirements by providing high throughput and minimum downtime while also providing power to PoE capable devices over the existing Ethernet network.
The 100-POE4 provides four pairs of two RJ-45 10/100BaseTX copper ports. Each pair consists of one RJ-45 port that connects to an existing network, and one RJ-45 PoE capable port that can send data and, up to 15.4 watts of power to a PoE capable device. The data on all four pairs are isolated from each other allowing for ease in upgrading non-PoE network infrastructures.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Input Current (mA) Max Input Voltage (VDC) Number of Ports
Availability Pricing
Red Lion 100-POE4 4 port 10/100BaseTX Industrial Ethernet PoE Midspan Power Injector, DIN-Rail EW-68586-14 Mfr # 100-POE4
COMPARE
Call For Price
Red Lion NTPS-48-5 DIN-Rail Power Supply, 5.0 Amp @ 48 VDC PoE PSU, and -HV Models EW-68586-15 Mfr # NTPS-48-5