This product is now discontinued, View Related Products

Nalidixic Acid

  • Assay - ≥98%
  • Heavy Metals (as Pb) - ≤0.002%
  • Melting Range - 225°-231°C
  • Loss on Drying at 105°C - ≤0.5%
Call For Price

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Fisher Chemical BP90825 Nalidixic Acid (25g) 8806850 EW-88068-50 Mfr # BP90825
COMPARE
389-08-2
Call For Price
Acros Organics AC169900250 Nalidixic acid 99.5% (25g) EW-88181-07 Mfr # AC169900250
COMPARE
389-08-2
Call For Price
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat