Neomycin Sulfate 26139

  • Purity - 90–95% Neomycin B
  • Potency - ≥630μg neomycin/mg
  • FTIR - Conforms to Standard
  • pH (3.3% aqueous solution) - 5–7.5
  • Loss on Drying - ≤8%
$90.50USD / Each

1 variations of this product are available.

See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Fisher BioReagents BP266925 Neomycin Sulfate (White Powder) (25g) CAS 1405-10-3 EW-88061-95 Mfr # BP266925
COMPARE
1405-10-3
$90.50USD / Each
Showing 1- 1 of 1

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat