This product is now discontinued, View Related Products
NIOSH Approved N-95 Particulate Respirators
NIOSH approved and CDC listed particulate respirator
  • Light weight, soft comfortable, latex-free material
  • Cushioning nose foam for comfort
  • Conforms to NIOSH 42CFR84
NIOSH approved particulate respirators protect against solid and liquid aerosols that do not contain oil. The ergonomic shape with welded straps gives you the perfect nose bridge seal. Optional exhalation valve keeps you cool.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Rating Description
Availability Pricing
Bal Supply NIOSH N95 Particulate Respirator, Nose Bridge Seal; 20/box EW-78901-24 Mfr # 05144
COMPARE
N-95 NIOSH N95 Particulate Respirator, Nose Bridge Seal
Call For Price
NIOSH N95 Particulate Respirator with valve, Nose Bridge Seal; 12/box EW-78901-25 Mfr # 15144
COMPARE
N-95 NIOSH N95 Particulate Respirator with valve, Nose Bridge Seal