NIOSH Approved N-99 Particulate Respirators
NIOSH approved and CDC listed particulate respirator
  • Light weight, soft comfortable, latex-free material
  • Cushioning nose foam for comfort
  • Conforms to NOISH 42CFR84
NIOSH approved particulate respirators protect against solid and liquid aerosols that do not contain oil. It features a flexible metal nose clip seal with stapled straps. Optional exhalation valve keeps you cool.
$19.50 - $21.50USD / Box of 12
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Rating Description
Availability Pricing
COMPARE
N-99 NIOSH N99 Particulate Respirator, Metal Nose Seal
$21.50USD / Box of 20
NIOSH N99 Particulate Respirator with valve, Metal Nose Seal; 12/box
Buy More Save More
EW-78901-27 Mfr # 1914
COMPARE
N-99 NIOSH N99 Particulate Respirator with valve, Metal Nose Seal
$19.50USD / Box of 12