Non-Spill Quick Disconnect Couplings
Less than 0.10mL spillage
  • One-hand operation
Virtually eliminate any loss of product with this leak-free design. Couplings are made of polypropylene with an EPDM O-ring for excellent chemical resistance. For a complete fitting, order one body and one insert. All bodies will mate with inserts to make a fitting that will work for multiple applications.
$14.10 - $479.00USD / Pkg of 25
29 variations of this product are available.
See All Product Options Below
29 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Compression Size / Tube Od (Port 1) (mm) Thread size NPT(M) (port 1) (in)
Availability Pricing
CPC (Colder) NS4D13006 Non-spill coupling body, polypropylene, 3/8" OD tubing compression fitting; 25/PK EW-31309-42 Mfr # NS4D13006
COMPARE
$452.00USD / Pkg of 25
CPC (Colder) NS4D10004BSPT Non-Spill Quick-Disconnect, PP Body, 1/4" BSPT(M), NS4; 25/PK EW-31309-43 Mfr # NS4D10004BSPT
COMPARE
$408.00USD / Pkg of 25
CPC (Colder) NS4D22002 Non-spill coupling insert, polypropylene, 1/8" ID tubing barb; 25/PK EW-31309-44 Mfr # NS4D22002
COMPARE
1/8
$309.00USD / Pkg of 25
CPC (Colder) NS6D40008 Non-Spill Quick-Disconnect, Panel-Mount PP Insert, 1/2" OD, NS6; 10/PK EW-31309-46 Mfr # NS6D40008
COMPARE
$256.00USD / Pkg of 10
CPC (Colder) NS4D42006 Non-spill panel-mount coupling insert, polypropylene, 3/8" ID tubing barb; 25/PK EW-31309-47 Mfr # NS4D42006
COMPARE
3/8
$479.00USD / Pkg of 25
CPC (Colder) NS4D20006 Non-spill coupling insert, polypropylene, 3/8" OD tubing compression fitting; 25/PK EW-31309-50 Mfr # NS4D20006
COMPARE
$349.00USD / Pkg of 25
CPC (Colder) NS6D42006 Non-Spill Quick-Disconnect Insert, 3/8" ID Tubing; 10/PK EW-31309-51 Mfr # NS6D42006
COMPARE
3/8
$219.00USD / Pkg of 10
CPC (Colder) NS6D17006 Non-Spill Quick-Disconnect Body, Polypropylene; 3/8" Hose Barb EW-31603-22 Mfr # NS6D17006
COMPARE
3/8
$26.50USD / Each
CPC (Colder) NS4D17004 Non-spill coupling body, polypropylene, 1/4" ID tubing barb EW-31603-23 Mfr # NS4D17004
COMPARE
1/4
$18.70USD / Each
CPC (Colder) NS6D17008 NS6 non-spill quick-disconnect body, polypropylene, 1/2" hose barb EW-31603-24 Mfr # NS6D17008
COMPARE
1/2
$26.50USD / Each
CPC (Colder) NS6D10008 Non-Spill Quick Disconnect Body, 1/2" NPT(M), Polypropylene, NS6 EW-31603-26 Mfr # NS6D10008
COMPARE
1/2
$25.50USD / Each
CPC (Colder) NS4D10004 Non-spill coupling body, polypropylene, 1/4" male NPT EW-31603-27 Mfr # NS4D10004
COMPARE
1/4
$18.10USD / Each
CPC (Colder) NS4D24004 Non-spill coupling insert, polypropylene, 1/4" male NPT EW-31603-29 Mfr # NS4D24004
COMPARE
1/4
$14.70USD / Each
CPC (Colder) NS4D17006 Non-Spill Quick-Disconnect, PP Body, 3/8" ID, NS4 31603-31
Always in Stock
EW-31603-31 Mfr # NS4D17006
COMPARE
3/8
$18.70USD / Each
CPC (Colder) NS6D22006 Non-Spill Quick-Disconnect Insert, Polypropylene; 3/8" Hose Barb EW-31603-32 Mfr # NS6D22006
COMPARE
3/8
$19.60USD / Each
CPC (Colder) NS4D24004BSPT Non-Spill Quick-Disconnect, PP Insert, 1/4" BSPT(M), NS4 EW-31603-33 Mfr # NS4D24004BSPT
COMPARE
$14.10USD / Each
CPC (Colder) NS6D22008 NS6 non-spill quick-disconnect insert, polypropylene, 1/2" hose barb
Always in Stock
EW-31603-34 Mfr # NS6D22008
COMPARE
1/2
$25.50USD / Each
CPC (Colder) NS4D22006 Coupling Insert, Non-Spill, 3/8" ID Tube, NS4
Always in Stock
EW-31603-35 Mfr # NS4D22006
COMPARE
3/8
$14.10USD / Each
CPC (Colder) NS6D24008 Non-Spill Quick Disconnect Insert, 1/2" NPT(M), Polypropylene, NS6 EW-31603-36 Mfr # NS6D24008
COMPARE
1/2
$19.70USD / Each
CPC (Colder) NS4D40006T Coupling Insert, Non-Spill, Panel Mount, 3/8" OD Tube EW-31605-13 Mfr # NS4D40006
COMPARE
$18.70USD / Each
Showing 1 - 20 of 29