Noshok 500 Series Precision Needle Valves
Soft-seat design
  • Soft seat design is 100% helium leak-tested to 1 x 10–4 mL/s at 200 psi
  • Designed with Delrin® acetal seat
  • Straight-through porting for bidirectional, high capacity flow and easy roddable cleaning
Series 500 needle valves are high quality instrument grade valves. They are the right choice when your application calls for positive shut-off or precise flow control. They work effectively with most gases and liquids.
$75.00 - $417.00USD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Inlet Port Outlet Port Wetted Materials
Availability Pricing
NOSHOK 502-FFC Needle Valve, Soft Seat Steel Female-Female 1/4"NPT EW-06292-35 Mfr # 502-FFC
COMPARE
1/4" NPT(F) 1/4" NPT(F) Stainless Steel
$75.00USD / Each
NOSHOK 502-FFS Needle Valve, Soft Seat 316 SS Female-Female 1/4"NPT EW-06292-36 Mfr # 502-FFS
COMPARE
1/4" NPT(F) 1/4" NPT(F) Stainless Steel
$170.00USD / Each
NOSHOK 502-MFC Needle Valve, Soft Seat Steel Male-Female 1/4"NPT EW-06292-37 Mfr # 502-MFC
COMPARE
1/4" NPT(M) 1/4" NPT(F) Stainless Steel
$83.50USD / Each
NOSHOK 502-MFS Needle Valve, Soft Seat 316 SS Male-Female 1/4"NPT EW-06292-38 Mfr # 502-MFS
COMPARE
1/4" NPT(M) 1/4" NPT(F) Stainless Steel
$212.00USD / Each
NOSHOK 503-FFC Needle Valve, Soft Seat Steel Male-Female 3/8" NPT EW-06292-39 Mfr # 503-FFC
COMPARE
3/8" NPT(M) 3/8" NPT(F) Stainless Steel
$82.50USD / Each
NOSHOK 503-FFS Needle Valve, Soft Seat Steel Male-Female 3/8" NPT EW-06292-40 Mfr # 503-FFS
COMPARE
3/8" NPT(M) 3/8" NPT(F) Stainless Steel
$208.00USD / Each
NOSHOK 50402-MFC Needle Valve, Soft Seat Steel 1/2" Male x 1/4" Female, NPT EW-06292-41 Mfr # 50402-MFC
COMPARE
1/2" NPT(M) 1/4" NPT(F) Stainless Steel
$91.50USD / Each
NOSHOK 504-FFC Needle Valve, Soft Seat Steel Female-Female 1/2"NPT EW-06292-42 Mfr # 504-FFC
COMPARE
1/2" NPT(F) 1/2" NPT(F) Stainless Steel
$82.50USD / Each
NOSHOK 504-FFS Needle Valve, Soft Seat 316 SS Female-Female 1/2"NPT EW-06292-43 Mfr # 504-FFS
COMPARE
1/2" NPT(F) 1/2" NPT(F) Stainless Steel
$208.00USD / Each
NOSHOK 504-MFC Needle Valve, Soft Seat Steel Male-Female 1/2"NPT EW-06292-44 Mfr # 504-MFC
COMPARE
1/2" NPT(M) 1/2" NPT(F) Stainless Steel
$91.50USD / Each
NOSHOK 504-MFS Needle Valve, Soft Seat 316 SS Male-Female 1/2"NPT EW-06292-45 Mfr # 504-MFS
COMPARE
1/2" NPT(M) 1/2" NPT(F) Stainless Steel
$235.00USD / Each
NOSHOK 508 FFC Needle Valve, Soft Seat Steel Female-Female 1" NPT EW-06292-46 Mfr # 508 FFC
COMPARE
1" NPT(F) 1" NPT(F) Stainless Steel
$417.00USD / Each
Showing 1- 12 of 12
Need Help with this product?
Call Us At1-847-549-7600
Live chat