NPS Emergency Response (Er) Accessories

Replace reagents included in the MERCSORB mercury spill kit
Replace the reagents within your 86464-00 mercury spill kit.
$25.50 - $72.50USD / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Spilfyter 520270 Replacement Amalgamation Powder, 270g EW-86464-50 Mfr # 520270
COMPARE
Replacement Amalgamation Powder, 270g
$25.50USD / Each
Spilfyter 523250 Replacement Mercury Indicator Powder, 250g EW-86464-52 Mfr # 523250
COMPARE
Replacement Mercury Indicator Powder, 250g
$72.50USD / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat