NSF-Certified Two-Way Solenoid Valves
Wetted materials fully compliant with NSF standards to ensure water and food safety
  • Valves create zero differential pressure
  • Integrated mounting holes for ease of installation or replacement
  • Low actuation wattage required, resulting in low energy consumption and operating cost
These two-way normally closed solenoid valves are used in sterilization, process, and vending of liquids, beverages, and food where NSF certification is required. High-quality virgin materials provide extended service life, and high pressure and temperature resistance The orifice food-grade sealing materials ensure compatibility with media and sealing for precise flow management.
$35.00 - $37.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Inlet Port Outlet Port
Availability Pricing
ODE 21JP1R1V23-T0 Solenoid Valve NSF-Certified 1/8" BSP(F) NC; PPS body; KFM Seal/PPS Seat EW-08617-94 Mfr # 21JP1R1V23-T0
COMPARE
1/8" NPT(F) 1/8" NPT(F)
$37.00USD / Each
ODE 21JPAR1V23-T0 NSF-Certified Two-Way Solenoid Valve, PPS; NC, 1/8"NPT(M), 24VAC EW-08617-96 Mfr # 21JPAR1V23-T0
COMPARE
1/8" NPT(M) 1/8" NPT(M)
$35.00USD / Each
Showing 1- 2 of 2
Need Help with this product?
Call Us At1-847-549-7600
Live chat