This product is now discontinued, View Related Products

o-Cresol 23362

Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Fisher Chemical C536250 o-Cresol (Laboratory) (250g) CAS 95-48-7 EW-88006-32 Mfr # C536250
COMPARE
95487 Laboratory
Call For Price
Acros Organics AC110550010 o-Cresol p.a. (1kg) CAS 95-48-7 (2-Hydroxytoluene, 2-Methylphenol) EW-88101-16 Mfr # AC110550010
COMPARE
95487
Call For Price
Acros Organics AC110555000 o-Cresol p.a. (500g) 8810118 EW-88101-18 Mfr # AC110555000
COMPARE
95-48-7
Call For Price
Acros Organics AC110560010 o-Cresol 98% (1kg) 8810119 EW-88101-19 Mfr # AC110560010
COMPARE
95-48-7
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat