Oakton® PC 2700 Benchtop pH/Conductivity Meters
Multiparameter benchtop meters are an all-in-one solution for laboratory research work
  • Advanced research-grade meter
  • Includes Ion concentration readings
Oakton PC 2700 meters feature pH auto-buffer recognition for both USA and NIST buffer sets, automatic or manual temperature compensation, and selectable temperature units. Previous calibration points, electrode slope and offset, and conductivity/TDS cell constants are stored in memory for quick and easy retrieval.
Meters offer an advanced user interface and advanced measurement capabilities. Meters meet GLP requirements—meters stamp all calibration and stored data with time/date. Nonvolatile memory stores up to 500 data sets to simplify documenting large volumes of pH, mV, ISE, conductivity, TDS, salinity, or resistivity measurements. Data is easily exported to a computer via RS-232 port. Order RS-232 and optional RS-232 to USB adapter separately.
$1,570.00 - $2,250.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Max Conductivity Max mV (mV) Max pH (pH)
Availability Pricing
COMPARE
0 - 500 mS 2000 20
$1,890.00USD / Each
COMPARE
0 - 500 mS 2000 20
$2,250.00USD / Each
COMPARE
0 - 500 mS 2000 20
$1,570.00USD / Each
COMPARE
0 - 500 mS 2000 20
$1,930.00USD / Each
Showing 1- 4 of 4
Need Help with this product?
Call Us At1-847-549-7600
Live chat