Oakton® pH 600 Waterproof Meters
Expanded range, resolution and accuracy, increased memory, and advanced communications!
  • No more excuses for missing data!
  • Out-of-date or unperformed calibrations are now things of the past!
  • Don't waste time looking for those cables!
  • Use in dirty and wet environments
These waterproof handheld meters feature a –2.00 to 20.00 pH range with 0.01 pH resolution. Combined with accuracies up to ±0.01 pH, these meters provide a new benchmark in our pH meter line. Store up to 500 data sets in the meter‘s memory. Time-and-date stamping meets Good Laboratory Practice (GLP) standards.
The pH 600 meters offer the latest in pH calibration capabilities including up to six calibration points, a user-settable “calibration due” alarm, and up to 15 buffer options with auto-buffer recognition of USA, NIST, DIN, and PWB standards.
The infrared (IrDA) wireless technology makes PC downloading more convenient and easier. Dual powerup allows you to use batteries while out in the field or the optional AC power adapter while in the lab.
As always, our waterproof meters are IP67 rated for protection against both dust and water, even with no probes attached.
ITEM(S) ON SALE
$753.10 - $1,255.50USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Waterproof Meter with pH probe
$878.05USD / Each
$1,033.00USD / Each
COMPARE
Waterproof Meter with pH probe with NIST-traceable Calibrations
$1,088.50USD / Each
COMPARE
pH 600 Waterproof Handheld Meter Only
$753.10USD / Each
$886.00USD / Each
COMPARE
Waterproof Meter Only with NIST-traceable Calibration
$954.75USD / Each
COMPARE
Waterproof Handheld Meter Kit
$1,039.12USD / Each
$1,222.50USD / Each
COMPARE
Waterproof Meter Kit with NIST-traceable Calibrations
$1,255.50USD / Each