This product is now discontinued, View Related Products
Oakton® Waterproof DO 600 Dissolved Oxygen Meters
Stores up to 500 data points with time-and-date stamp for GLP compliance
  • Programmable “calibration due” date for increased data integrity
  • Wirelessly download data via IrDA or RS-232 (LED) port
These top-of-the-line waterproof meters offer single or multiparameter measurement capabilities to meet all of your testing needs. The IP67-rated housing allows use in dirty and wet environments without harm to the meter. Store up to 500 data points with manual or auto datalogging and download to computer or printer with the infrared (IrDA) wireless technology — no need for additional cords. User-selectable “calibration due” and set point alarms, automatic or manual standards recognition, and single- or multiple-point calibration provide flexibility in meeting your data quality objectives. The large, backlit graphic LCD shows the measured value and units, temperature, time, battery life, and menu options.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Max Dissolved Oxygen Min Dissolved Oxygen
Availability Pricing
COMPARE
0 to 50 ppm, 0 to 600% saturation
Call For Price
COMPARE
0 to 50 ppm, 0 to 600% saturation
Call For Price
COMPARE
0 to 50 ppm, 0 to 600% saturation
Call For Price
COMPARE
0 to 50 ppm, 0 to 600% saturation
Call For Price
COMPARE
0 to 50 ppm, 0 to 600% saturation
Call For Price
COMPARE
0 to 50 ppm, 0 to 600% saturation