This product is now discontinued, View Related Products

Ohaus Champ/CKW Washdown Industrial Scales

Easy-to-clean design with stainless steel construction
  • 1:3000 certified resolution
  • All stainless steel construction
  • Feature IP66/NEMA 4X-rated housing and IP67-rated load cell
  • Removable stainless steel platform with extended skirt
  • Checkweighing methods: positive, negative, zero/net
  • Easily monitor check weighing results via three-color bar graph with audible and display alerts
  • Statistical functions including accumulation, average, and standard deviation
Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity (kg) Capacity (lbs) Power (VAC)
Availability Pricing
Ohaus CKW3R55 Champ/CKW Washdown Industrial Scales, 3kg/6Lb, 120V, 1: 5000 Resolution EW-01007-18 Mfr # CKW3R55
COMPARE
3 6 100 to 240
Call For Price
Ohaus CKW6R55 Champ/Ckw Washdown Industrial Scales, 6kg/15Lb, 120V, 1: 7500 Resolution EW-01007-22 Mfr # CKW6R55
COMPARE
6 15 100 to 240
Call For Price
Ohaus CKW15L55 Champ/CKW Washdown Industrial Scales, 15kg/30lb, 120V, 1: 7500 Resolution EW-01007-24 Mfr # CKW15L55
COMPARE
15 30 100 to 240
Call For Price
Ohaus CKW3R55 Champ/CKW Washdown Industrial Scales, 30kg/60lb, 120V, 1: 6000 Resolution EW-01007-25 Mfr # CKW30L55
COMPARE
3 60 100 to 240
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat