Ohaus Defender W-Series Washdown High Resolution Industrial Scale Base Only

  • Removable 304 stainless steel weighing pans offer quick cleaning
  • Stainless steel frame
  • Durability, rigidity and structural integrity
  • IP-67 OIML R60 stainless steel load celll
These durable rectangular bases feature welded tubular steel construction for industrial applications. Rugged, adjustable feet help prevent slippage on wet or uneven surfaces
$891.00 - $2,022.00USD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Readability (g)
Availability Pricing
Ohaus D10WR Defender W-Series Washdown Base 10kg 12" x 12" EW-90452-91 Mfr # D10WR
COMPARE
10 25 1
$891.00USD / Each
Ohaus D25WR Defender W-Series Washdown Base 25kg 12" x 12" EW-90452-92 Mfr # D25WR
COMPARE
25 50 2
$891.00USD / Each
Ohaus D50WL Defender W-Series Washdown Base 50kg 18" x 18" EW-90452-93 Mfr # D50WL
COMPARE
50 100 5
$956.00USD / Each
Ohaus D100WL Defender W-Series Washdown Base 100kg 18" x 18" EW-90452-94 Mfr # D100WL
COMPARE
100 250 10
$1,322.00USD / Each
Ohaus D250WX Defender W-Series Washdown Base 250kg 24" x 24" EW-90452-95 Mfr # D250WX
COMPARE
250 500 20
$2,022.00USD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat