Ohaus Harvard Trip Mechanical Balances 62445
Two-pan design is ideal for comparative weighing
  • Comparative weighing to determine the difference in mass between two objects
  • Select models have a tare beam to accommodate containers up to 225g
  • Magnetic damping speeds up weigh times
  • Built-in sliding masses
ITEM(S) ON SALE
$377.72 - $421.80USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Readability (g) Description
Availability Pricing
Ohaus 1450-SD Harvard Triple Balance, 2000g x 0.1g, Single Beam, Stainless Steel Plates EW-11110-19 Mfr # 1450-SD
COMPARE
2000 0.1 Harvard Triple Balance, 2000g x 0.1g, Single Beam, Stainless Steel Plates
$377.72USD / Each
$497.00USD / Each
Ohaus 1550-SD Harvard Triple Balance, 2000g x 0.1g, Two Beam, Stainless Steel Plates EW-11110-21 Mfr # 1560-SD
COMPARE
2000 0.1 Harvard Triple Balance, 2000g x 0.1g, Two Beam, Stainless Steel Plates
$403.56USD / Each
$531.00USD / Each
Ohaus 1560-SD Harvard Triple Balance, 2000g x 0.1g, Two Beam, Stainless Steel Plates, Tare EW-11110-22 Mfr # 1560-SD
COMPARE
2000 0.1 Harvard Triple Balance, 2000g x 0.1g, Two Beam, Stainless Steel Plates, Tare
$421.80USD / Each
$555.00USD / Each