Ohaus MB Moisture Balances
Versatile and rugged design for continuous operation in food and beverage, quality control, environmental, and many other applications
  • Choice of infrared or halogen heat source and programmable operation satisfy a wider variety of applications
  • Durable, compact construction ensures dependable, long-term operation within minimum benchtop space
  • Straightforward operation speeds up routine testing applications and accommodates operators of all skill levels
Ohaus MB Series moisture analyzers are the quick, effective, and affordable solution to measure moisture content. These moisture analyzers cover a wider variety of weighing applications in the pharmaceutical, chemical and research industries with excellent repeatability properties for consistent, accurate testing.
ITEM(S) ON SALE
$3,269.72USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Heater Type Readability (g)
Availability Pricing
Ohaus MB23 MB Moisture Determination Balance, Mb23 110g x 0.01g 120V EW-01002-81 Mfr # MB23
COMPARE
110 Infrared 0.01
$3,269.72USD / Each
$3,802.00USD / Each
Ohaus MB23 MB Moisture Determination Balance, Mb23 110g x 0.01g 230V EW-01002-86 Mfr # MB23-230VAC
COMPARE
110 Infrared 0.001
$3,269.72USD / Each
$3,802.00USD / Each