This product is now discontinued, View Related Products
OHAUS TAJ Series Jewelry / Precious Metals Balances
Ensure fair trade—comply with FTC guidelines
  • Accurate weighing and counting promote good business practices
  • Add USB or RS-232 connectivity (order separately) to document and print data
  • Security features include rear security loop, integral transportation lock, and calibration lock-out
  • AC power or battery (four AA, not included) operation increase versatility
The OHAUS TAJ Series was designed for precision portable weighing for a wide range of jewelry applications.
From the Carat Series with high precision and a quick-open draftshield to the Gold Series for
higher capacity weighing of precious metals and stones, the TAJ is a great value for your basic weighing
needs.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Capacity (g) Description
Availability Pricing
Ohaus TAJ203 Carat Balance, 200 ct x 0.005 ct EW-11335-71 Mfr # TAJ203
COMPARE
40 Carat Balance, 200 ct x 0.005 ct
Call For Price
Ohaus TAJ202 Gold Balance, 200 g x 0.01 g EW-11335-72 Mfr # TAJ202
COMPARE
200 Gold Balance, 200 g x 0.01 g
Call For Price
Ohaus TAJ501 Gold Balance, 500 g x 0.1 g EW-11335-73 Mfr # TAJ501
COMPARE
500 Gold Balance, 500 g x 0.1 g
Call For Price
Ohaus TAJ602  Gold Balance, 600 g x 0.01 g EW-11335-74 Mfr # TAJ602
COMPARE
600 Gold Balance, 600 g x 0.01 g
Call For Price
Ohaus TAJ4001  Gold Balance, 4000 g x 0.1 g EW-11335-75 Mfr # TAJ4001
COMPARE
4000 Gold Balance, 4000 g x 0.1 g
Call For Price
Ohaus TAJ6000 Gold Balance, 6000 g x 1 g EW-11335-76 Mfr # TAJ6000
COMPARE
6000 Gold Balance, 6000 g x 1 g