Ohaus Valor 1000 Compact Bench Scales
Economical, multipurpose weighing in a compact precision scale
  • THIS SCALE IS NOT WASHDOWN RATED
  • Included internal rechargeable battery offers portability and up to 100 hours of battery life
  • Perform routine weighing, accumulation, and checkweighing tasks with 4-button keypad and simple, menu-driven operation
  • Straightforward, intuitive display indicators including over/accept/under, center of zero, stability, net, sum, weighing units, and battery charge status
  • Stainless steel weighing platform can be removed for easy cleanup
Ohaus Valor 1000 compact precision scales are an economical choice for your basic weighing needs. Standard software includes checkweighing mode, accumulation mode and multiple units of measure. Each scale includes a long lasting internal rechargeable battery for portability and convenience.
ITEM(S) ON SALE
$217.08 - $244.24USD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Power (VAC)
Availability Pricing
Ohaus V11P15T Valor 1000  Compact Industrial Scale, 15 kg X 2 g EW-01030-04 Mfr # V11P15T
COMPARE
15 33 120
$244.24USD / Each
$284.00USD / Each
Ohaus V11P30T Valor 1000 Compact Industrial Scale, 30 kg X 5 g EW-01030-05 Mfr # V11P30T
COMPARE
30 66 120
$244.24USD / Each
$284.00USD / Each
Ohaus V11P3T Valor 1000 Compact Industrial Scale, 3 kg X 0.5 g EW-01030-06 Mfr # V11P3T
COMPARE
3 6.6 120
$244.24USD / Each
$284.00USD / Each
Ohaus V11P6T Valor 1000 Compact Industrial Scale, 6 kg X 1 g EW-01030-07 Mfr # V11P6T
COMPARE
6 13 120
$244.24USD / Each
$284.00USD / Each
Ohaus V11P3 Valor 1000 Compact Industrial Scale, 3, 000g, x 0.5g 115 V EW-11011-31 Mfr # V11P3
COMPARE
3 6.5 115
$230.48USD / Each
$268.00USD / Each
Ohaus V11P3 Valor 1000 Compact Industrial Scale, 3, 000g x 0.5g, 230 V EW-11011-32 Mfr # V11P3
COMPARE
3 6.5 230
$217.08USD / Each
$268.00USD / Each
Ohaus V11P6 Valor 1000 Compact Industrial Scale, 6, 000g x 1g, 115 V EW-11011-33 Mfr # V11P6
COMPARE
6 13 115
$230.48USD / Each
$268.00USD / Each
Ohaus V11P6 Valor 1000 Compact Industrial Scale, 6, 000g x 1g, 230 V EW-11011-34 Mfr # V11P6
COMPARE
6 13 230
$217.08USD / Each
$268.00USD / Each
Ohaus V11P15 Valor 1000 Compact Industrial Scale, 15, 000g x 2g, 115 V EW-11011-35 Mfr # V11P15
COMPARE
15 33 115
$230.48USD / Each
$268.00USD / Each
Ohaus V11P15 Valor 1000 Compact Industrial Scale, 15, 000g x 2g, 230 V EW-11011-36 Mfr # V11P15
COMPARE
15 33 230
$230.48USD / Each
$268.00USD / Each
Ohaus V11P30 Valor 1000 Compact Industrial Scale, 30, 000g x 5g, 115 V
Always in Stock
EW-11011-37 Mfr # V11P30
COMPARE
30 66 115
$230.48USD / Each
$268.00USD / Each
Ohaus V11P30 Valor 1000 Compact Industrial Scale, 30, 000g x 5g, 230 V EW-11011-38 Mfr # 83998129
COMPARE
30 66 230
$230.48USD / Each
$268.00USD / Each
Showing 1- 12 of 12