Cole-Parmer® Essentials Single-Channel Adjustable-Volume Pipettor
Ergonomic finger grip for comfortable use
  • Easy to operate with its ergonomic, lightweight push-button design
  • Forward and reverse pipetting modes
  • Drop of the pipette is color-coded by volume for easy identification
  • Volume is displayed through handgrip window for quick glance convenience
  • Adjustable-volume pipettors feature click-stop volume settings
These pipettors are offered in a wide-range of adjustable volumes to lend to a variety of precise liquid dispensing needs. They are fully autoclavable.
$165.00USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Increments (µL) Max Volume (µL) Min Volume (µL)
Availability Pricing
COMPARE
0.01 2.5 0.1
$165.00USD / Each
COMPARE
0.1 10 0.5
$165.00USD / Each
COMPARE
0.5 50 5
$165.00USD / Each
COMPARE
0.5 100 10
$165.00USD / Each
COMPARE
1 200 20
$165.00USD / Each
COMPARE
5 1000 100
$165.00USD / Each
COMPARE
50 5000 500
$165.00USD / Each
COMPARE
100 10000 2000
$165.00USD / Each
Showing 1- 8 of 8