One-Way Luer Check Valves
Valves feature low priming volume, ease of removing air bubbles during priming and operate in any spatial orientation. Valves are manufactured DEHP-and latex-free silicone diaphragms. Materials meet USP CLass VI and ISO 10993 specifications.
$11.75 - $14.50USD / Pkg of 10
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Connection Type-Port 1 Connection Type-Port 2 Material
Availability Pricing
COMPARE
Female luer Male luer lock SAN
$14.50USD / Pkg of 10
COMPARE
Female luer Male luer lock SAN
$11.75USD / Pkg of 10
Showing 1- 2 of 2
Need Help with this product?
Call Us At1-847-549-7600
Live chat