Monarch Instrument Optical Tachometer Reflective Tape
Cut and adhere to surface being monitored
All optical tachometers require a reflective surface to operate—use reflective tape to create this surface.
$25.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Max Measuring Distance (in) Max RPM Range (rpm) Memory
Availability Pricing
Monarch MI153209 Roll of Reflective Tape for use with Optical Tachometers, 5 ft long and 1/2" wide
Always in Stock
EW-08210-62 Mfr # 6180-070
COMPARE
$25.00USD / Each