Orbital-Genie Benchtop Shakers
Available in three platform versions for any type of container Can be used in cold room, incubator, and CO2 environments
  • Fully programmable
These shakers allow you to create any mixing or shaking action, from a smooth vortex to sloshing aeration. Shakers feature variable speeds including orbit reverse, programmable acceleration and deceleration rates, and a built-in timer/alarm ranging from 1 minute to 99 hours. Ramping feature ensures smooth acceleration to desired speed while avoiding potential spills.
$3,440.00 - $4,730.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
Scientific Industries Genie SI-1500 Orbital Shaker with Ratcheting Clamps; 120V EW-51820-82 Mfr # SI-1500
COMPARE
120
$4,730.00USD / Each
Scientific Industries Genie SI-1502 Orbital Shaker with Ratcheting Clamps; 230V EW-51820-86 Mfr # SI-1502
COMPARE
230
$4,490.00USD / Each
Scientific Industries Genie SI-1600 Orbital Shaker for Standard Flask Clamps; 120V EW-51820-88 Mfr # SI-1600
COMPARE
120
$3,440.00USD / Each
Scientific Industries Genie SI-1602 Orbital Shaker for Standard Flask Clamps; 230V EW-51820-92 Mfr # SI-1602
COMPARE
230
$3,440.00USD / Each
Scientific Industries Genie SI-1700 Orbital Shaker with Adhering Mat; 120V EW-51820-94 Mfr # SI-1700
COMPARE
120
$3,850.00USD / Each
Scientific Industries Genie SI-1702 Orbital Shaker with Adhering Mat; 230V EW-51820-98 Mfr # SI-1702
COMPARE
230
$3,750.00USD / Each
Showing 1- 6 of 6
Need Help with this product?
Call Us At1-847-549-7600
Live chat